Learn More
MSC Ansell Work Gloves, High-Visibility, Synthetic Leather Palm, Slip-On, 1 Pair
Catalog No. 5000788319
Encompass
These high-visibility work gloves are designed for flexible, versatile hand protection. They offer improved durability with a grip-enhancing synthetic leather palm, increased comfort from premium mesh fabric, and practical features like a slip-on cuff and touchscreen compatibility.
  • Split palm with premium synthetic leather for heightened durability.
  • Top-of-hand premium mesh fabric for enhanced comfort and flexibility.
  • Slip-on elastic cuff for easy-to-use everyday personal protection.
  • Touchscreen-compatible thumb, index, and middle finger for unhindered hand protection.
  • High-visibility external surface and reflective logo for enhanced visibility.
Catalog No. 50-007-88319 Supplier MSC Supplier No. 59626051
These high-visibility work gloves are designed for flexible, versatile hand protection. They offer improved durability with a grip-enhancing synthetic leather palm, increased comfort from premium mesh fabric, and practical features like a slip-on cuff and touchscreen compatibility.
  • Split palm with premium synthetic leather for heightened durability.
  • Top-of-hand premium mesh fabric for enhanced comfort and flexibility.
  • Slip-on elastic cuff for easy-to-use everyday personal protection.
  • Touchscreen-compatible thumb, index, and middle finger for unhindered hand protection.
  • High-visibility external surface and reflective logo for enhanced visibility.