Learn More
Medchemexpress LLC Anserine-d4 | 1201658-81-2 | 99.4% | 244.28 | 1 MG
SDP
Catalog No. 5000436556
Encompass_Preferred
Anserine-d4 is a deuterium-labeled version of Anserine, which is a naturally occurring, orally active Histidine-containing dipeptide found in the skeletal muscle of vertebrates. Anserine acts as a biochemical buffer, chelator, antioxidant, and anti-glycation agent, and is known to improve memory functions.
  • Deuterium-labeled compound for research applications.
  • Functions as a biochemical buffer.
  • Acts as a chelator.
  • Serves as an antioxidant.
  • Exhibits anti-glycation properties.
  • May improve memory functions.
Catalog No. 50-004-36556 Supplier Medchemexpress LLC Supplier No. HY113354S1MG
Anserine-d4 is a deuterium-labeled version of Anserine, which is a naturally occurring, orally active Histidine-containing dipeptide found in the skeletal muscle of vertebrates. Anserine acts as a biochemical buffer, chelator, antioxidant, and anti-glycation agent, and is known to improve memory functions.
  • Deuterium-labeled compound for research applications.
  • Functions as a biochemical buffer.
  • Acts as a chelator.
  • Serves as an antioxidant.
  • Exhibits anti-glycation properties.
  • May improve memory functions.