Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ansofaxine hydrochloride | 916918-84-8 | 417.97 | 1 ML
SDP

Catalog No. 5000448538
Encompass_Preferred

Ansofaxine hydrochloride (LY03005) is a triple reuptake inhibitor that targets serotonin, dopamine, and norepinephrine. It is intended for research use only.

  • Inhibits serotonin reuptake (IC50: 723 nM)
  • Inhibits dopamine reuptake (IC50: 491 nM)
  • Inhibits norepinephrine reuptake (IC50: 763 nM)
  • Purity of 99.44%
  • Molecular weight of 417.97

Catalog No. 50-004-48538 Supplier Medchemexpress LLC Supplier No. HYU0009610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ansofaxine hydrochloride (LY03005) is a triple reuptake inhibitor that targets serotonin, dopamine, and norepinephrine. It is intended for research use only.

  • Inhibits serotonin reuptake (IC50: 723 nM)
  • Inhibits dopamine reuptake (IC50: 491 nM)
  • Inhibits norepinephrine reuptake (IC50: 763 nM)
  • Purity of 99.44%
  • Molecular weight of 417.97

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.