Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ansofaxine hydrochloride | 916918-84-8 | 417.97 | 25 MG
SDP

Catalog No. 5000448014
Encompass_Preferred

Ansofaxine hydrochloride is a triple reuptake inhibitor designed to inhibit serotonin, dopamine, and norepinephrine reuptake. It demonstrates IC50 values of 723 nM, 491 nM, and 763 nM for these reuptake processes, respectively. This compound is intended for research use only.

  • Triple reuptake inhibitor
  • Inhibits serotonin, dopamine, and norepinephrine reuptake
  • Intended for research use only
  • Appears as a solid
  • Color ranges from white to off-white
  • Recommended storage for solid is 4°C, sealed and away from moisture
  • For solutions, store at -80°C for up to 6 months, or -20°C for 1 month

Catalog No. 50-004-48014 Supplier Medchemexpress LLC Supplier No. 25MGHYU00096
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ansofaxine hydrochloride is a triple reuptake inhibitor designed to inhibit serotonin, dopamine, and norepinephrine reuptake. It demonstrates IC50 values of 723 nM, 491 nM, and 763 nM for these reuptake processes, respectively. This compound is intended for research use only.

  • Triple reuptake inhibitor
  • Inhibits serotonin, dopamine, and norepinephrine reuptake
  • Intended for research use only
  • Appears as a solid
  • Color ranges from white to off-white
  • Recommended storage for solid is 4°C, sealed and away from moisture
  • For solutions, store at -80°C for up to 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.