Learn More
Medchemexpress LLC Ansofaxine hydrochloride | 916918-84-8 | 417.97 | 50 MG
SDP
Catalog No. 5000448929
Encompass_Preferred
Ansofaxine hydrochloride is a triple reuptake inhibitor that inhibits serotonin, dopamine, and norepinephrine reuptake. In vivo studies show it rapidly penetrates the rat striatum, converts into desvenlafaxine, and increases 5-HT, dopamine, and norepinephrine levels, ultimately reducing immobility time.
  • Inhibits serotonin, dopamine, and norepinephrine reuptake
  • Rapidly penetrates rat striatum
  • Increases 5-HT, dopamine, and norepinephrine levels
  • Reduces immobility time
Catalog No. 50-004-48929 Supplier Medchemexpress LLC Supplier No. HYU0009650MG
Ansofaxine hydrochloride is a triple reuptake inhibitor that inhibits serotonin, dopamine, and norepinephrine reuptake. In vivo studies show it rapidly penetrates the rat striatum, converts into desvenlafaxine, and increases 5-HT, dopamine, and norepinephrine levels, ultimately reducing immobility time.
  • Inhibits serotonin, dopamine, and norepinephrine reuptake
  • Rapidly penetrates rat striatum
  • Increases 5-HT, dopamine, and norepinephrine levels
  • Reduces immobility time