Learn More
New England Biolabs, Inc. Antarctic Thermolabile UDG – 100 units
SDP
Catalog No. 50591114
Encompass
Antarctic Thermolabile UDG (Uracil- DNA Glycosylase) catalyzes the release of free uracil from uracil-containing single-stranded or double-stranded DNA. The resulting abasic sites are susceptible to hydrolytic cleavage at elevated temperature and high pH. This enzyme is sensitive to heat and can be rapidly and completely inactivated at temperatures above 50C.
Catalog No. 50-591-114 Supplier New England Biolabs, Inc. Supplier No. M0372S
Antarctic Thermolabile UDG (Uracil- DNA Glycosylase) catalyzes the release of free uracil from uracil-containing single-stranded or double-stranded DNA. The resulting abasic sites are susceptible to hydrolytic cleavage at elevated temperature and high pH. This enzyme is sensitive to heat and can be rapidly and completely inactivated at temperatures above 50C.