Learn More
ABclonal Technology ANT1/ANT2/ANT3/ANT4 Rabbit pAb
SDP
Catalog No. 5000106562
Encompass_Preferred
ANTs are the mitochondrial carrier subfamily of solute carrier proteins. ANT protein functions as a gated pore that translocates ADP from the cytoplasm into the mitochondrial matrix and ATP from the mitochondrial matrix into the cytoplasm. The protein forms a homodimer embedded in the inner mitochondria membrane.
Catalog No. 50-001-06562 Supplier ABclonal Technology Supplier No. A159881000UL
ANTs are the mitochondrial carrier subfamily of solute carrier proteins. ANT protein functions as a gated pore that translocates ADP from the cytoplasm into the mitochondrial matrix and ATP from the mitochondrial matrix into the cytoplasm. The protein forms a homodimer embedded in the inner mitochondria membrane.