Learn More
ABclonal Technology ANT2/SLC25A5 Rabbit pAb
SDP
Supplier:  ABclonal Technology A156391000UL
Encompass_Preferred
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the cytoplasm into the mitochondrial matrix and ATP from the mitochondrial matrix into the cytoplasm. The protein forms a homodimer embedded in the inner mitochondria membrane. Suppressed expression of this gene has been shown to induce apoptosis and inhibit tumor growth. The human genome contains several non-transcribed pseudogenes of this gene.
Catalog No. 50-001-07551