Learn More
Medchemexpress LLC Ant308 (TFA) | 99.6% | 3467.10 | 5 MG
SDP
Catalog No. 5000393407
Encompass_Preferred
ANT308 TFA is a vasoactive intestinal polypeptide (VIP receptor) antagonist. It significantly enhances the activation and proliferation of T cells, inhibits migration and metastasis, and induces apoptosis of melanoma tumor cells by inhibiting VIP-VPAC2 signaling and reducing the expression of MCAM and N-cadherin. It can be used for the study of acute myeloid leukemia (AML) and uveal melanoma (UVM).
  • Reduces the viability of specific cell lines
  • Decreases the proportion of cells in the S phase
  • Induces apoptosis in certain cell types
  • Inhibits cell migration in multiple cell lines
  • Reduces MCAM and N-cadherin expression
  • Reduces the number and size of liver metastases in animal models
  • Shows a trend toward reducing tumor volume in primary tumor sites in animal models
Catalog No. 50-003-93407 Supplier Medchemexpress LLC Supplier No. 5MGHYP11288A
ANT308 TFA is a vasoactive intestinal polypeptide (VIP receptor) antagonist. It significantly enhances the activation and proliferation of T cells, inhibits migration and metastasis, and induces apoptosis of melanoma tumor cells by inhibiting VIP-VPAC2 signaling and reducing the expression of MCAM and N-cadherin. It can be used for the study of acute myeloid leukemia (AML) and uveal melanoma (UVM).
  • Reduces the viability of specific cell lines
  • Decreases the proportion of cells in the S phase
  • Induces apoptosis in certain cell types
  • Inhibits cell migration in multiple cell lines
  • Reduces MCAM and N-cadherin expression
  • Reduces the number and size of liver metastases in animal models
  • Shows a trend toward reducing tumor volume in primary tumor sites in animal models