Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antalarmin hydrochloride | 220953-69-5 | MFCD05664721 | 99.9% | 415.01 g/mol | C24H35ClN4 | 1 ML
SDP

Catalog No. 5000245054
Encompass_Preferred

Antalarmin hydrochloride is a non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used in research to suppress CRH-induced ACTH secretion and to study stress- and inflammation-related responses in animal models.

  • Selective CRHR1 antagonist with reported Ki ≈ 1 nM.
  • CAS number 220953-69-5.
  • Molecular formula C24H35ClN4, MW 415.01 g/mol.
  • Available as a solid or as a 10 mM solution in DMSO (1 mL).
  • High purity (~99.9%) suitable for analytical and in vivo preparations.
  • Recommended storage: solid at -20°C; in solvent store at -80°C for long term.

Catalog No. 50-002-45054 Supplier Medchemexpress LLC Supplier No. HY10337710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antalarmin hydrochloride is a non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used in research to suppress CRH-induced ACTH secretion and to study stress- and inflammation-related responses in animal models.

  • Selective CRHR1 antagonist with reported Ki ≈ 1 nM.
  • CAS number 220953-69-5.
  • Molecular formula C24H35ClN4, MW 415.01 g/mol.
  • Available as a solid or as a 10 mM solution in DMSO (1 mL).
  • High purity (~99.9%) suitable for analytical and in vivo preparations.
  • Recommended storage: solid at -20°C; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.