Learn More
Medchemexpress LLC Antalarmin hydrochloride | 220953-69-5 | MFCD05664721 | 99.9% | 415.01 g/mol | C24H35ClN4 | 100 MG
SDP
Catalog No. 5000244752
Encompass_Preferred
Antalarmin hydrochloride is the hydrochloride salt of antalarmin, a non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist commonly used in preclinical research. It is supplied as a high-purity research chemical suitable for both in vitro and in vivo studies investigating stress-related pathways and CRHR1 pharmacology.
  • High purity suitable for analytical and preclinical use.
  • Orally active CRHR1 antagonist for in vivo studies.
  • Chemical formula C24H35ClN4 and molecular weight 415.01 g/mol.
  • Available in small research quantities for dose-ranging experiments.
Catalog No. 50-002-44752 Supplier Medchemexpress LLC Supplier No. HY103377100MG
Antalarmin hydrochloride is the hydrochloride salt of antalarmin, a non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist commonly used in preclinical research. It is supplied as a high-purity research chemical suitable for both in vitro and in vivo studies investigating stress-related pathways and CRHR1 pharmacology.
  • High purity suitable for analytical and preclinical use.
  • Orally active CRHR1 antagonist for in vivo studies.
  • Chemical formula C24H35ClN4 and molecular weight 415.01 g/mol.
  • Available in small research quantities for dose-ranging experiments.