Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antalarmin hydrochloride | 220953-69-5 | MFCD05664721 | 99.9% | 415.01 g·mol⁻¹ | C24H35ClN4 | 5 MG
SDP

Catalog No. 5000245158
Encompass_Preferred

Antalarmin hydrochloride is the hydrochloride salt of antalarmin, a non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used in research to block CRH-induced ACTH secretion and study stress- and anxiety-related pathways. It is supplied as a solid powder with high purity and provided for in vitro and in vivo experimental applications.

  • Non-peptide CRHR1 antagonist for research use.
  • Purity: 99.87%.
  • Molecular weight: 415.01 g·mol⁻¹.
  • Physical form: white to light yellow solid powder.
  • Storage: store solid at -20°C; in solution, -80°C for up to 6 months.
  • Solubility: soluble in DMSO (≈125 mg/mL), DMF, and ethanol; follow vehicle recommendations for in vivo use.
  • Supplied in a 5 mg pack for small-scale research studies.

Catalog No. 50-002-45158 Supplier Medchemexpress LLC Supplier No. HY1033775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antalarmin hydrochloride is the hydrochloride salt of antalarmin, a non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used in research to block CRH-induced ACTH secretion and study stress- and anxiety-related pathways. It is supplied as a solid powder with high purity and provided for in vitro and in vivo experimental applications.

  • Non-peptide CRHR1 antagonist for research use.
  • Purity: 99.87%.
  • Molecular weight: 415.01 g·mol⁻¹.
  • Physical form: white to light yellow solid powder.
  • Storage: store solid at -20°C; in solution, -80°C for up to 6 months.
  • Solubility: soluble in DMSO (≈125 mg/mL), DMF, and ethanol; follow vehicle recommendations for in vivo use.
  • Supplied in a 5 mg pack for small-scale research studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.