Learn More
New England Biolabs, Inc. Antarctic Phosphatase – 5000 units
SDP
Catalog No. 50811709
Encompass
Antarctic Phosphatase (AnP) is a heat labile alkaline phosphatase purified from a recombinant source. AnP nonspecifically catalyzes the dephosphorylation of 5' and 3' ends of DNA and RNA phosphomonoesters. Also, AnP Hydrolyses ribo-, as well as deoxyribonucleoside triphosphates (NTPs and dNTPs). AnP is useful in many molecular biology applications such as the removal of phosphorylated ends of DNA and RNA for subsequent use in cloning or end-labeling of probes. In cloning, dephosphorylation prevents religation of linearized plasmid DNA. The enzyme acts on 5' protruding, 5' recessed, and blunt ends. AnP may also be used to degrade unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis. AnP is completely and irreversibly inactivated by heating at 80C for 2 minutes, thereby making removal of AnP prior to ligation or end-labeling unnecessary.
Catalog No. 50-811-709 Supplier New England Biolabs, Inc. Supplier No. M0289L
Antarctic Phosphatase (AnP) is a heat labile alkaline phosphatase purified from a recombinant source. AnP nonspecifically catalyzes the dephosphorylation of 5' and 3' ends of DNA and RNA phosphomonoesters. Also, AnP Hydrolyses ribo-, as well as deoxyribonucleoside triphosphates (NTPs and dNTPs). AnP is useful in many molecular biology applications such as the removal of phosphorylated ends of DNA and RNA for subsequent use in cloning or end-labeling of probes. In cloning, dephosphorylation prevents religation of linearized plasmid DNA. The enzyme acts on 5' protruding, 5' recessed, and blunt ends. AnP may also be used to degrade unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis. AnP is completely and irreversibly inactivated by heating at 80C for 2 minutes, thereby making removal of AnP prior to ligation or end-labeling unnecessary.