Learn More
New England Biolabs, Inc. Antarctic Thermolabile UDG – 500 units
SDP
Catalog No. 501526390
Encompass
Antarctic Thermolabile UDG (Uracil- DNA Glycosylase) catalyzes the release of free uracil from uracil-containing single-stranded or double-stranded DNA. The resulting abasic sites are susceptible to hydrolytic cleavage at elevated temperature and high pH. This enzyme is sensitive to heat and can be rapidly and completely inactivated at temperatures above 50C.
Catalog No. 50-152-6390 Supplier New England Biolabs, Inc. Supplier No. M0372L
Antarctic Thermolabile UDG (Uracil- DNA Glycosylase) catalyzes the release of free uracil from uracil-containing single-stranded or double-stranded DNA. The resulting abasic sites are susceptible to hydrolytic cleavage at elevated temperature and high pH. This enzyme is sensitive to heat and can be rapidly and completely inactivated at temperatures above 50C.