Learn More
Apexbio Technology LLC Antazoline HCl 2508-72-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089101
Encompass_Preferred
Antazoline hydrochloride (CAS 2508-72-7) is a first-generation antihistamine that acts as a competitive antagonist at the histamine H1 receptor By occupying the H1 binding site Antazoline HCl prevents endogenous histamine from eliciting its effects thereby inhibiting downstream signaling pathways associated with allergic responses This compound is frequently utilized in pharmacological studies to investigate histaminergic signaling allergy mechanisms and the physiological role of H1 receptor modulation in various in vitro and in vivo models
Catalog No. 50-000-89101 Supplier Apexbio Technology LLC Supplier No. B16645.1
Antazoline hydrochloride (CAS 2508-72-7) is a first-generation antihistamine that acts as a competitive antagonist at the histamine H1 receptor By occupying the H1 binding site Antazoline HCl prevents endogenous histamine from eliciting its effects thereby inhibiting downstream signaling pathways associated with allergic responses This compound is frequently utilized in pharmacological studies to investigate histaminergic signaling allergy mechanisms and the physiological role of H1 receptor modulation in various in vitro and in vivo models