Learn More
Medchemexpress LLC Antazoline hydrochloride | 2508-72-7 | 5 G
SDP
Catalog No. 5000413972
Encompass_Preferred
Antazoline hydrochloride is a histamine H1 receptor antagonist with anticholinergic and antiviral properties. It prevents histamine from acting on target cells through reversible competition and exerts an antiallergic effect. It can reduce HBV DNA in a dose-dependent manner and has a significant inhibitory effect on HBV DNA in Huh7 cells. It also has anti-arrhythmic effects and can be studied for cardiovascular diseases and HBV.
  • Histamine H1 receptor antagonist
  • Exhibits anticholinergic and antiviral properties
  • Prevents histamine from acting on target cells
  • Exerts an antiallergic effect
  • Reduces HBV DNA in a dose-dependent manner
  • Has a significant inhibitory effect on HBV DNA in Huh7 cells
  • Shows anti-arrhythmic effects in acute myocardial infarctions
  • Can be studied for cardiovascular diseases and HBV
Catalog No. 50-004-13972 Supplier Medchemexpress LLC Supplier No. HYB10675G
Antazoline hydrochloride is a histamine H1 receptor antagonist with anticholinergic and antiviral properties. It prevents histamine from acting on target cells through reversible competition and exerts an antiallergic effect. It can reduce HBV DNA in a dose-dependent manner and has a significant inhibitory effect on HBV DNA in Huh7 cells. It also has anti-arrhythmic effects and can be studied for cardiovascular diseases and HBV.
  • Histamine H1 receptor antagonist
  • Exhibits anticholinergic and antiviral properties
  • Prevents histamine from acting on target cells
  • Exerts an antiallergic effect
  • Reduces HBV DNA in a dose-dependent manner
  • Has a significant inhibitory effect on HBV DNA in Huh7 cells
  • Shows anti-arrhythmic effects in acute myocardial infarctions
  • Can be studied for cardiovascular diseases and HBV