Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antazoline Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. A213225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A213225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A213225G Supplier TCI America Supplier No. A213225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2508-72-7
Molecular Formula C17H20ClN3
Molecular Weight (g/mol) 301.82
MDL Number MFCD00058145
InChI Key SWKDMSRRIBZZAY-UHFFFAOYSA-N
Synonym antazoline hydrochloride, antazoline hcl, fenazolina, histazine, 2-n-benzylanilinomethyl-2-imidazoline hydrochloride, antazolinium chloratum, antistine hydrochloride, phenazoline hydrochloride, unii-fp8q8f72jh
PubChem CID 17275
IUPAC Name hydrogen N-benzyl-N-[(4,5-dihydro-1H-imidazol-2-yl)methyl]aniline chloride
SMILES [H+].[Cl-].C(N(CC1=CC=CC=C1)C1=CC=CC=C1)C1=NCCN1

Specifications

Melting Point 238°C
Color White-Yellow
Quantity 25 g
Formula Weight 301.82
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Antazoline Hydrochloride
RTECSNumber NJ2150000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.