Learn More
ACROBiosystems Antibody Internalization Detection Reagent, 100 tests
Catalog No. NC3723312
Encompass_Preferred
Antibody Internalization Detection Reagent labels human IgG Fc with a pH-sensitive red fluorescent dye to enable real-time monitoring of antibody endocytosis in live cells. It forms a stable fluorescent complex rapidly and is compatible with flow cytometry and fluorescence imaging.
  • Labels human IgG Fc with a pH-sensitive red fluorescent dye.
  • Enables real-time monitoring of antibody endocytosis in live cells.
  • Compatible with flow cytometry and fluorescence imaging.
  • High signal-to-noise ratio with bright fluorescence in acidic compartments.
  • Rapid labeling, complete in about 10 minutes.
  • Lyophilized for long-term storage; protect from light and avoid repeated freeze-thaw cycles.
Catalog No. NC3723312 Supplier ACROBiosystems Supplier No. IGGPZF2001100TESTS
Antibody Internalization Detection Reagent labels human IgG Fc with a pH-sensitive red fluorescent dye to enable real-time monitoring of antibody endocytosis in live cells. It forms a stable fluorescent complex rapidly and is compatible with flow cytometry and fluorescence imaging.
  • Labels human IgG Fc with a pH-sensitive red fluorescent dye.
  • Enables real-time monitoring of antibody endocytosis in live cells.
  • Compatible with flow cytometry and fluorescence imaging.
  • High signal-to-noise ratio with bright fluorescence in acidic compartments.
  • Rapid labeling, complete in about 10 minutes.
  • Lyophilized for long-term storage; protect from light and avoid repeated freeze-thaw cycles.