Learn More
TARGETMOL CHEMICALS INC antcin A 1MG
SDP
Catalog No. 502877303
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. antcin A is a steroid-like phytochemical isolated from the fruiting bodies of a precious edible mushroom Antrodia cinnamomea with strong anti-inflammatory and anti-tumor effects. Purity 96.43%
Catalog No. 50-287-7303 Supplier TARGETMOL CHEMICALS INC Supplier No. TN70991MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. antcin A is a steroid-like phytochemical isolated from the fruiting bodies of a precious edible mushroom Antrodia cinnamomea with strong anti-inflammatory and anti-tumor effects. Purity 96.43%