Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Analyzer Corporation Kit, Furnace, 120V/240V 13" Fiber
SDP

Catalog No. NC2127665
Encompass

Assy, Furnace, 120V/240V 13" Fiber EOS 12-31-2026

This item is not returnable. View return policy
Catalog No. NC2127665 Supplier Petroleum Analyzer Corporation Supplier No. 103291
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Assy, Furnace, 120V/240V 13" Fiber EOS 12-31-2026

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.