Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Anteobind(Tm) Biosensor

Catalog No. APLSC010 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APLSC010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APLSC010 Supplier Sigma Aldrich Supplier No. APLSC01010ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

AnteoBind Biosensor is a ready-to-use activation reagent specifically developed for binding biomolecules onto planar surfaces including polystyrenes, certain cyclic olefin copolymers, microarray slides, silicon oxides, titanium oxides, ceramics and gold surfaces (excluding gold colloids in suspension). AnteoBind Biosensor can modify the strength of binding, wettability and surface charge of base materials commonly used in biosensor applications. By using AnteoBind surface coatings, it is possible to control protein binding and improve the stability of bound biomolecules across a range of surfaces used in biosensors.AnteoBind Biosensor has many advantages over methods such as EDC. AnteoBind is water based and replaces the dry chemicals you would use with the EDC method. Using AnteoBind instead of NHS/EDC chemistry can save up to 3-4 hours of lab time. Experiments have shown increased stability and functionality for antibodies and a decreased background. AnteoBind improves reproducibility between experiments by providing uniformity between surfaces.Activation and coupling of biosensor planar surfacesAnteobind is a trademark of AnteoTech Ltd

Specifications

Quantity 1 g
Synonym (S)-2-(tert-butoxycarbonylamino)-3-(2-methoxyphenyl)propanoic acid
Molecular Weight (g/mol) 295.33 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.