Learn More
Crescent Chemical Co Inc ANTHANTHRENE SOLN 1 ML
SDP
Supplier:  Crescent Chemical Co Inc PSXA20515000AL
Encompass_Preferred
Anthanthrene 100 µg/mL in Acetonitrile Polycyc. arom. hydrocarb. (WS Lsg) CAS 191-26-4 Molecular Weight - 276.3307 Storage - RT Catalog Num - DRE-XA20515000AL chemical
Catalog No. 50-809-02