Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anthra[1,2-c]thiadiazole-6,11-dione | 4219-52-7 | 98.0% | 10MG
SDP

Catalog No. 5000233610
Encompass_Preferred

NSC745885 is an anti-tumor research compound that down-regulates EZH2 via proteasome-mediated degradation and shows selective cytotoxicity against multiple cancer cell lines. It is supplied as a high-purity research reagent for laboratory use.

  • High purity (98.0%) suitable for in vitro research.
  • Molecular weight 266.27 g/mol; formula C14H6N2O2S.
  • Available in small research pack sizes, including 10 mg.
  • Soluble in DMSO; often formulated as a suspension.
  • Store powder at -20°C; in solution store at -80°C for long-term.

Catalog No. 50-002-33610 Supplier Medchemexpress LLC Supplier No. HY11919810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NSC745885 is an anti-tumor research compound that down-regulates EZH2 via proteasome-mediated degradation and shows selective cytotoxicity against multiple cancer cell lines. It is supplied as a high-purity research reagent for laboratory use.

  • High purity (98.0%) suitable for in vitro research.
  • Molecular weight 266.27 g/mol; formula C14H6N2O2S.
  • Available in small research pack sizes, including 10 mg.
  • Soluble in DMSO; often formulated as a suspension.
  • Store powder at -20°C; in solution store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.