Learn More
Medchemexpress LLC Anthra[1,2-c]thiadiazole-6,11-dione | 4219-52-7 | 98.0% | 5 MG
SDP
Catalog No. 5000231892
Encompass_Preferred
NSC745885 is a small-molecule anti-tumor agent reported to down-regulate EZH2 through proteasome-mediated degradation. It has demonstrated selective cytotoxicity against multiple cancer cell lines and is used in preclinical cellular research of bladder and oral squamous cell carcinoma.
  • Down-regulates EZH2 via proteasome-mediated degradation.
  • Exhibits selective cytotoxicity toward malignant cell lines.
  • Supplied as a solid with high purity suitable for research.
  • Stable when stored at low temperatures according to handling guidance.
  • Available in small research quantities for laboratory studies.
Catalog No. 50-002-31892 Supplier Medchemexpress LLC Supplier No. HY1191985MG
NSC745885 is a small-molecule anti-tumor agent reported to down-regulate EZH2 through proteasome-mediated degradation. It has demonstrated selective cytotoxicity against multiple cancer cell lines and is used in preclinical cellular research of bladder and oral squamous cell carcinoma.
  • Down-regulates EZH2 via proteasome-mediated degradation.
  • Exhibits selective cytotoxicity toward malignant cell lines.
  • Supplied as a solid with high purity suitable for research.
  • Stable when stored at low temperatures according to handling guidance.
  • Available in small research quantities for laboratory studies.