Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anthra[1,2-c]thiadiazole-6,11-dione | 4219-52-7 | 98.0% | 50 MG
SDP

Catalog No. 5000233929
Encompass_Preferred

NSC745885 (anthra[1,2-c]thiadiazole-6,11-dione) is a small-molecule anti-tumor research compound supplied as a solid powder with high purity for in vitro and preclinical studies.

  • Small-molecule anti-tumor scaffold (anthra[1,2-c]thiadiazole-6,11-dione).
  • CAS 4219-52-7.
  • Purity 98.0%.
  • Molecular formula C14H6N2O2S; molecular weight 266.27 g/mol.
  • Solid powder; available in 5 mg-100 mg sizes.
  • Storage: powder stable at -20°C for long-term; in solvent store at -80°C for short-term.

Catalog No. 50-002-33929 Supplier Medchemexpress LLC Supplier No. HY11919850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

NSC745885 (anthra[1,2-c]thiadiazole-6,11-dione) is a small-molecule anti-tumor research compound supplied as a solid powder with high purity for in vitro and preclinical studies.

  • Small-molecule anti-tumor scaffold (anthra[1,2-c]thiadiazole-6,11-dione).
  • CAS 4219-52-7.
  • Purity 98.0%.
  • Molecular formula C14H6N2O2S; molecular weight 266.27 g/mol.
  • Solid powder; available in 5 mg-100 mg sizes.
  • Storage: powder stable at -20°C for long-term; in solvent store at -80°C for short-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.