Learn More
Medchemexpress LLC Anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetrone, 2,9-bis(4-ethoxyphenyl)- | 24108-89-2 | 98.01% | 5 G
SDP
Catalog No. 5000411074
Encompass_Preferred
C.I. Pigment red 123 is a significant colorant used to add or change the color of various items. It remains chemically stable and largely unaffected by the carrier or medium it is incorporated into. This pigment finds wide application across numerous industries due to its versatility and stability.
  • Used as a colorant in diverse applications
  • Chemically stable and largely unaffected by carrier medium
  • Suitable for use in textiles, medicine, food, cosmetics, plastics, paint, ink, photography, and paper industries
  • For research use only
Catalog No. 50-004-11074 Supplier Medchemexpress LLC Supplier No. HYD05315G
C.I. Pigment red 123 is a significant colorant used to add or change the color of various items. It remains chemically stable and largely unaffected by the carrier or medium it is incorporated into. This pigment finds wide application across numerous industries due to its versatility and stability.
  • Used as a colorant in diverse applications
  • Chemically stable and largely unaffected by carrier medium
  • Suitable for use in textiles, medicine, food, cosmetics, plastics, paint, ink, photography, and paper industries
  • For research use only