Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Anthracene

Catalog No. 501859109
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-9109 100 g
50-185-9110 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-9109 Supplier Sigma Aldrich Supplier No. A89200100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Anthracene
TRUSTED_SUSTAINABILITY

Specifications

CAS 120-12-7
Melting Point 210°C to 215°C (lit.)
Boiling Point 340°C (lit.)
Molecular Formula C14H10
Linear Formula C14H10
MDL Number MFCD00001240
Quantity 100 g
Synonym Anthraxcene; Paranaphthalene
Molecular Weight (g/mol) 178.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.