Learn More
Medchemexpress LLC Anthracene-9-carboxylic acid | 723-62-6 | MFCD00001257 | 222.24 g/mol | C15H10O2 | 1 ML
SDP
Catalog No. 5000212534
Encompass_Preferred
Anthracene-9-carboxylic acid is an anthracene derivative supplied as a 10 mM solution in DMSO and as solid material. It is used in electrophysiology and cellular studies to block and identify Ca2+-activated Cl- currents (CaCCs). Chemical identifiers: CAS 723-62-6; formula C15H10O2; molecular weight 222.24 g/mol.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • Available as solid for synthesis and analytical use.
  • Acts as a blocker and identifier of Ca2+-activated Cl- currents in cell studies.
  • Well-characterized chemical properties for reproducible experimental use.
Catalog No. 50-002-12534 Supplier Medchemexpress LLC Supplier No. HY10132910MM 1ML
Anthracene-9-carboxylic acid is an anthracene derivative supplied as a 10 mM solution in DMSO and as solid material. It is used in electrophysiology and cellular studies to block and identify Ca2+-activated Cl- currents (CaCCs). Chemical identifiers: CAS 723-62-6; formula C15H10O2; molecular weight 222.24 g/mol.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • Available as solid for synthesis and analytical use.
  • Acts as a blocker and identifier of Ca2+-activated Cl- currents in cell studies.
  • Well-characterized chemical properties for reproducible experimental use.