Learn More
Medchemexpress LLC Anthracene-9-carboxylic acid | 723-62-6 | MFCD00001257 | 222.24 g/mol | C15H10O2 | 500 MG
SDP
Catalog No. 5000211387
Encompass_Preferred
Anthracene-9-carboxylic acid is an anthracene derivative supplied as an analytical standard for research use. It is commonly used as a chloride current blocker in electrophysiology and as a reference material in analytical testing.
  • Analytical standard grade for research applications.
  • Molecular formula C15H10O2; molecular weight 222.24 g/mol.
  • Used to block Ca2+-activated chloride currents in electrophysiology assays.
  • Suitable as a reference standard for chromatographic and spectroscopic analyses.
  • Available in multiple pack sizes, including 500 mg.
Catalog No. 50-002-11387 Supplier Medchemexpress LLC Supplier No. HY101329500MG
Anthracene-9-carboxylic acid is an anthracene derivative supplied as an analytical standard for research use. It is commonly used as a chloride current blocker in electrophysiology and as a reference material in analytical testing.
  • Analytical standard grade for research applications.
  • Molecular formula C15H10O2; molecular weight 222.24 g/mol.
  • Used to block Ca2+-activated chloride currents in electrophysiology assays.
  • Suitable as a reference standard for chromatographic and spectroscopic analyses.
  • Available in multiple pack sizes, including 500 mg.