Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anthracene-9-carboxylic acid-d9 | 1219803-78-7 | 98.9% | 231.29 g/mol | C15HD9O2 | 1 MG
SDP

Catalog No. 5000211508
Encompass_Preferred

Anthracene-9-carboxylic acid-d9 is the deuterium-labeled analog of anthracene-9-carboxylic acid supplied as a high-purity solid for use as an isotopic standard or tracer in analytical and research applications. It is suitable for mass spectrometry calibration, quantification, and tracer studies where a stable isotope internal standard is required.

  • Isotopically labeled for accurate internal standardization in mass spectrometry.
  • High purity (98.9%) for reliable analytical results.
  • Small 1 mg solid quantity ideal for trace-level standards and calibration.
  • CAS number 1219803-78-7 for unambiguous chemical identification.
  • Molecular weight 231.29 g/mol and formula C15HD9O2 for reference.

Catalog No. 50-002-11508 Supplier Medchemexpress LLC Supplier No. HY101329S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anthracene-9-carboxylic acid-d9 is the deuterium-labeled analog of anthracene-9-carboxylic acid supplied as a high-purity solid for use as an isotopic standard or tracer in analytical and research applications. It is suitable for mass spectrometry calibration, quantification, and tracer studies where a stable isotope internal standard is required.

  • Isotopically labeled for accurate internal standardization in mass spectrometry.
  • High purity (98.9%) for reliable analytical results.
  • Small 1 mg solid quantity ideal for trace-level standards and calibration.
  • CAS number 1219803-78-7 for unambiguous chemical identification.
  • Molecular weight 231.29 g/mol and formula C15HD9O2 for reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.