Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthracene-d10 Solution, 1000μg/mL in methylene chloride, Ultra Scientific

Catalog No. USIST1101
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-IST-1101 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-IST-1101 Supplier Agilent Technologies Supplier No. IST1101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1719-06-8 , 75-09-2
Molecular Formula C14H10
Molecular Weight (g/mol) 188.295
InChI Key MWPLVEDNUUSJAV-LHNTUAQVSA-N
PubChem CID 80291
IUPAC Name 1,2,3,4,5,6,7,8,9,10-decadeuterioanthracene
SMILES C1=CC=C2C=C3C=CC=CC3=CC2=C1

Specifications

CAS Max % 99.93, 0.07
Analyte or Component Names Methylene chloride,anthracene-d10
Element Methylene Chloride, Anthracene-d10
Solvent or Matrix Methylene Chloride
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Anthracene-d10 Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.