Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthracene, MilliporeSigma™ Supelco™

Catalog No. 111019413 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9413 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9413 Supplier MilliporeSigma Supelco Supplier No. 31581250MG
Add to Cart
Add to Cart

Anthracene is a low-molecular weight polycyclic aromatic hydrocarbon (PAH). It acts as starting material for the production of anthraquinone.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 1004°F
CAS 120-12-7
Vapor Pressure 1 mmHg (145°C)
Vapor Density 6.15 (vs. air)
Physical Form Neat
Linear Formula C14H10
MDL Number MFCD00001240
Melting Point 210°C to 215°C
Boiling Point 340°C
Synonym Anthraxcene; Paranaphthalene
Solubility Soluble in hydronaphthalenes
Type Analytical Standard
Quantity 250 mg
Formula Weight 178.23
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.