Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthracene 97.0+%, TCI America™
SDP

Catalog No. A0495100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0495100G 100 g
A049525G 25 g
A0495500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0495100G Supplier TCI America Supplier No. A0495100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 218°C
Boiling Point 342°C
Quantity 100 g
UN Number 3077
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Anthracene
EINECSNumber (4)-0683
RTECSNumber CA9350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.