Learn More
Medchemexpress LLC HY-W031757 250mg Medchemexpress, Anthraquinone-2-carboxylic acid CAS:117-78-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW031757250MG
Encompass_Preferred
Medchemexpress, HY-W031757 250mg Anthraquinone-2-carboxylic acid CAS:117-78-2 Anthraquinone-2-carboxylic acid is a major anthraquinone isolated from Brazilian taheebo, with anti-inflammatory activity and antinociceptive. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3447