Learn More
Medchemexpress LLC Anthraquinone-2-carboxylic acid | 117-78-2 | MFCD00001231 | 98.0% | 252.22 g/mol | C15H8O4 | 10 G
SDP
Catalog No. 5000301728
Encompass_Preferred
Anthraquinone-2-carboxylic acid is an anthraquinone derivative (CAS 117-78-2) supplied as an off-white to light yellow solid for research use. It has reported anti-inflammatory and antinociceptive activity and is provided at ~98.0% purity. Solubility in DMSO is 25 mg/mL (ultrasonic recommended). Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • High purity suitable for in vitro research.
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO at 25 mg/mL; ultrasonic recommended.
  • Long-term stability when stored under recommended conditions.
  • Provided for research use only; not validated for clinical applications.
Catalog No. 50-003-01728 Supplier Medchemexpress LLC Supplier No. HYW03175710G
Anthraquinone-2-carboxylic acid is an anthraquinone derivative (CAS 117-78-2) supplied as an off-white to light yellow solid for research use. It has reported anti-inflammatory and antinociceptive activity and is provided at ~98.0% purity. Solubility in DMSO is 25 mg/mL (ultrasonic recommended). Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • High purity suitable for in vitro research.
  • Off-white to light yellow solid appearance.
  • Soluble in DMSO at 25 mg/mL; ultrasonic recommended.
  • Long-term stability when stored under recommended conditions.
  • Provided for research use only; not validated for clinical applications.