Learn More
Medchemexpress LLC Anthraquinone-2-carboxylic acid | 117-78-2 | MFCD00001231 | 98.0% | 252.22 g/mol | C15H8O4 | 25 G
SDP
Catalog No. 5000304020
Encompass_Preferred
Anthraquinone-2-carboxylic acid is a research-grade anthraquinone derivative (C15H8O4, MW 252.22 g/mol) reported as a major constituent of Brazilian taheebo. Manufacturer literature describes anti-inflammatory and antinociceptive activity. Supplied as solid and as a 10 mM solution in DMSO, this compound is intended for laboratory research use only.
  • Small-molecule anthraquinone derivative with reported anti-inflammatory activity.
  • Also reported to have antinociceptive effects in biological studies.
  • Available as solid and as a 10 mM solution in DMSO for flexible use.
  • High purity suitable for research applications.
  • Provided with product datasheet information including molecular formula and molecular weight.
Catalog No. 50-003-04020 Supplier Medchemexpress LLC Supplier No. 25GHYW031757
Anthraquinone-2-carboxylic acid is a research-grade anthraquinone derivative (C15H8O4, MW 252.22 g/mol) reported as a major constituent of Brazilian taheebo. Manufacturer literature describes anti-inflammatory and antinociceptive activity. Supplied as solid and as a 10 mM solution in DMSO, this compound is intended for laboratory research use only.
  • Small-molecule anthraquinone derivative with reported anti-inflammatory activity.
  • Also reported to have antinociceptive effects in biological studies.
  • Available as solid and as a 10 mM solution in DMSO for flexible use.
  • High purity suitable for research applications.
  • Provided with product datasheet information including molecular formula and molecular weight.