Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Anthraquinone-2-sulfonic acid sodium salt monohydrate

Catalog No. 1232421KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-324-21KG 1 kg
12-324-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-324-21KG Supplier Sigma Aldrich Supplier No. 1232421KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Anthraquinone-2-sulfonic acid sodium salt monohydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate.anthraquinone-2-sulfonic acid sodium salt monohydrate was used to investigate the effects of reactant concentration and synthesis parameters on the electrical conductivity of polypyrrole coated polyethyleneterephthalate fabrics
  • It has been used as dopant in the preparation of polypyrrole films by electrochemical polymerization

Specifications

CAS 153277-35-1
Molecular Formula C14H7NaO5S -+ H2O
Linear Formula C14H7NaO5S -+ H2O
MDL Number MFCD00149068
Quantity 1 kg
Synonym 9,10-Dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt monohydrate; Sodium anthraquinone-2-sulfonate
Molecular Weight (g/mol) 328.27
Percent Purity 97%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.