Learn More
Medchemexpress LLC Anthraquinone | 84-65-1 | 208.21 | 25 G
SDP
Catalog No. 5000389474
Encompass_Preferred
Anthraquinone is a chemical precursor used in dye formation and agrochemicals. It is found in various organisms and exhibits a range of biological activities, including anticancer, anti-inflammatory, and antioxidant properties. It can also inhibit the topoisomerase II enzyme, leading to cell death via apoptosis.
  • Precursor for dye formation and agrochemicals
  • Exhibits anticancer, anti-inflammatory, and antioxidant properties
  • Affects primary plant metabolism by inhibiting energy transfer in photosynthesis
  • Can intercalate into DNA and inhibit topoisomerase II enzyme
  • Leads to cell death via apoptosis
Catalog No. 50-003-89474 Supplier Medchemexpress LLC Supplier No. 25GHYN0354
Anthraquinone is a chemical precursor used in dye formation and agrochemicals. It is found in various organisms and exhibits a range of biological activities, including anticancer, anti-inflammatory, and antioxidant properties. It can also inhibit the topoisomerase II enzyme, leading to cell death via apoptosis.
  • Precursor for dye formation and agrochemicals
  • Exhibits anticancer, anti-inflammatory, and antioxidant properties
  • Affects primary plant metabolism by inhibiting energy transfer in photosynthesis
  • Can intercalate into DNA and inhibit topoisomerase II enzyme
  • Leads to cell death via apoptosis