Learn More
Medchemexpress LLC Anthraquinone | 84-65-1 | 208.21 | 250 G
SDP
Catalog No. 5000389519
Encompass_Preferred
Anthraquinone is a compound that serves as a precursor for dye formation and agrochemicals. It is naturally present in various organisms such as bacteria, fungi, plants, and certain animals. This compound exhibits a range of biological activities and plays a role in plant metabolism.
  • Exhibits anticancer, anti-inflammatory, diuretic, antiarthritic, antifungal, antibacterial, antimalarial, and antioxidant properties.
  • Influences the electron transport chain in plants, inhibiting energy transfer during photosynthesis.
  • Can intercalate into DNA and inhibit the topoisomerase II (topo II) enzyme, inducing cell death via apoptosis.
Catalog No. 50-003-89519 Supplier Medchemexpress LLC Supplier No. 250GHYN0354
Anthraquinone is a compound that serves as a precursor for dye formation and agrochemicals. It is naturally present in various organisms such as bacteria, fungi, plants, and certain animals. This compound exhibits a range of biological activities and plays a role in plant metabolism.
  • Exhibits anticancer, anti-inflammatory, diuretic, antiarthritic, antifungal, antibacterial, antimalarial, and antioxidant properties.
  • Influences the electron transport chain in plants, inhibiting energy transfer during photosynthesis.
  • Can intercalate into DNA and inhibit the topoisomerase II (topo II) enzyme, inducing cell death via apoptosis.