Learn More
Medchemexpress LLC Anthraquinone-d8 | 10439-39-1 | 216.26 | 5 MG
SDP
Catalog No. 5000390079
Encompass_Preferred
Anthraquinone-d8 is a deuterium-labeled compound, primarily utilized as a precursor for dye formation. It serves as a valuable tracer and internal standard in quantitative analysis, making it suitable for various research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-90079 Supplier Medchemexpress LLC Supplier No. HYN0354S5MG
Anthraquinone-d8 is a deuterium-labeled compound, primarily utilized as a precursor for dye formation. It serves as a valuable tracer and internal standard in quantitative analysis, making it suitable for various research applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs