Learn More
Enzo Life Sciences Anthrax Lethal Factor monoclonal antibody (LF2) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905621100
Encompass_Preferred
Clone: LF2. Host: Mouse. Isotype: IgG1. Immunogen: Anthrax lethal factor. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Bacillus anthracis. Applications: ELISA, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.01% sodium azide. Concentration 1mg/ml.
Catalog No. 50-200-6969