Learn More
Cusabio Technology LLC anthrax toxin receptor 1
Catalog No. 5000051081
Encompass_Preferred
The Human Anthrax Toxin Receptor 1 (ANTXR1) ELISA Kit is designed for the quantitative measurement of ANTXR1 in serum plasma and tissue homogenates using a sandwich immunoassay format This kit detects ANTXR1 with a sensitivity of 15 6 pg/mL and a dynamic detection range from 62 5 pg/mL to 4000 pg/mL ensuring precise quantification across biological samples The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours making it efficient for routine laboratory use Detection is performed at 450 nm and the kit includes optimized reagents for robust performance Its sandwich-based design ensures high specificity while the broad detection range accommodates diverse experimental needs This reliable and standardized tool supports research applications requiring accurate ANTXR1 level assessment in multiple sample types
Catalog No. 50-000-51081 Supplier Cusabio Technology LLC Supplier No. CSBEL001832HU
The Human Anthrax Toxin Receptor 1 (ANTXR1) ELISA Kit is designed for the quantitative measurement of ANTXR1 in serum plasma and tissue homogenates using a sandwich immunoassay format This kit detects ANTXR1 with a sensitivity of 15 6 pg/mL and a dynamic detection range from 62 5 pg/mL to 4000 pg/mL ensuring precise quantification across biological samples The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours making it efficient for routine laboratory use Detection is performed at 450 nm and the kit includes optimized reagents for robust performance Its sandwich-based design ensures high specificity while the broad detection range accommodates diverse experimental needs This reliable and standardized tool supports research applications requiring accurate ANTXR1 level assessment in multiple sample types