Learn More
Abcam Anthrone, carbohydrate determination reagent, 25G
Catalog No. 502704667
Encompass_Preferred
MW 194.23 g/mol. Tricyclic aromatic hydrocarbon used in the colorometric determination of carbohydrates.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4667 Supplier Abcam Supplier No. AB14638525G
MW 194.23 g/mol. Tricyclic aromatic hydrocarbon used in the colorometric determination of carbohydrates.
The product is subject to the following: Abcam Restricted Use Statement