Learn More
ECM Bioscience δ1-Catenin (Tyr-228), phospho-specific Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM3561
Encompass_Preferred
Clone M356 was generated from a phospho-δ1-Catenin (Tyr-228) synthetic peptide containing amino acid residues around tyrosine 228 of human δ1-Catenin. This peptide sequence is highly conserved in rat and mouse δ1-Catenin. Accession #O60716. The antibody detects a 110 kDa* protein corresponding to the molecular mass of δ1-Catenin on SDS-PAGE immunoblots of human A431 and HUVEC cells treated with pervanadate. This antibody does not detect this band after alkaline phosphatase treatment. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = pp120 Src substrate, p120
Catalog No. 50-206-8663