Learn More
Aviva Systems Biology Corp Fam172a Antibody - N-terminal region (ARP37639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against 1110033M05Rik. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of mouse 1110033M05Rik. Alternate names: 53-E6, pEN87, AF064782, 1110033M05Rik, 2610318O14Rik, 9430037D06RikApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-53840