Learn More
Aviva Systems Biology Corp Use1 Antibody - N-terminal region (ARP37352_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37352_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against 2010315L10RIK. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of mouse 2010315L10RIK. Alternate names: D12, Ed2, p31, Q-snare, AV002165, 2010315L10Rik, 5730403H22RikApplicationIHC, WB. Species reactivity: Mouse, Rat
Catalog No. 50-014-53818