Learn More
Aviva Systems Biology Corp Mri1 Antibody - C-terminal region (ARP37363_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37363_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against 2410018C20RIK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2410018C20RikApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53819