Learn More
AdipoGen anti-4-Hydroxy-2-nonenal [4-HNE], mAb (HNEJ-2)
SDP
Supplier:  AdipoGen JAIMHN020P
Encompass_Preferred
anti-4-Hydroxy-2-nonenal [4-HNE], mAb (HNEJ-2), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes 4-HNE-lysine, 4-HNE-histidine and 4-HNE cysteine adduct. Has much higher affinity for the 4-HNE-histidine adduct than 4-HNE-lysine or 4-HNE cysteine adduct. Shows almost negligible reactivity with proteins that were treated with other aldehydes, such as 2-nonenal, 2-hexenal, 1-hexanal, 4-hydroxy-2-hexenal, formaldehyde, or glutaraldehyde. Lyophilized. Contains 50mM Tris-buffered saline (TBS). Reactive oxygen species (ROS) are involved in lipid peroxidation inside the living bodies, and membrane lipids are one of the major targets of ROS. During the peroxidation process, a variery of aldehydes are formed.
Catalog No. 50-101-5742