Learn More
Cosmo Bio Usa Inc Anti 4R-Tau Antibody, 50 uL, rabbit polyclonal, recognizes 4R tau isoforms, unlabeled, for Western Blotting and Immunohistochemistry, liquid form
SDP
Supplier:  Cosmo Bio Usa Inc CACTIP4RTP01
Encompass
The Anti 4R-Tau Antibody is a rabbit polyclonal antibody designed to specifically recognize 4R tau isoforms that are significant in Alzheimer's Disease. This antibody is generated from rabbits immunized with a synthetic peptide and is validated for use in Western Blotting (WB) and Immunohistochemistry (IHC), making it a reliable tool for examining tau species across various neurodegenerative diseases.
  • Recognizes 4R tau isoforms, important for distinguishing tau pathologies in neurodegenerative conditions.
  • Displays cross-reactivity with human, mouse, and rat samples, accommodating diverse research applications.
  • Validated for Western Blotting and Immunohistochemistry (paraffin sections) for effective protein detection and tissue localization.
Catalog No. NC0902643