Learn More
FUJIFILM BIOSCIENCES INC Anti 4R-Tau Monoclonal Antibod
Supplier:  FUJIFILM BIOSCIENCES INC 01326591
Encompass_Preferred
Tau is a microtubule-associated protein mainly expressed in neurons in the central nervous system and regulates stabilization of microtubules In the brain of Alzheimers disease patients neurofibrillary tangles are formed which are composed of accumulated phosphorylated forms of Tau The degree of these tangles correlates to the severity of dementia Based on this finding Tau has been investigated to find out the etiology and to discover drugs for Alzheimers disease Tau is classified into 3R-Tau and 4R-Tau which has 3 and 4 microtubules associated domains respectively This product is a mouse monoclonal antibody specific for the 4-repeat isoform of tau 4R-Tau Specific for 4R isoform of Tau
Catalog No. 50-004-50138